1-Formylpiperazine 90%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Piperazinecarboxaldehyde, 1-Piperazinecarbaldehyde

38849-0250 38849-1000
TS38849-0250EA 59 USD
TS38849-0250 TS38849-1000
1-Formylpiperazine 90%
1-Formylpiperazine
Formula: C₅H₁₀N₂O
MW: 114.15 g/mol
Boiling Pt: 105…107 °C (1 mmHg)
Density: 1.1 g/cm³
Flash Pt: 102 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005963
CAS Number: 7755-92-2
EINECS: 231-813-0

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5090 to 1.5130 (20°C, 589 nm)
GC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR