(S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, (2S)-(-)-2-Methoxy-2-phenyl-3,3,3-trifluoropropanoic acid

16823-0010 16823-2500
TS16823-0010EA 155 USD
TS16823-0010 TS16823-2500
(S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid 98%
(S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid
Formula: C₁₀H₉F₃O₃
MW: 234.17 g/mol
Boiling Pt: 116…118 °C (1.5 mmHg)
Density: 1.303 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064200
CAS Number: 17257-71-5
EINECS: 241-292-1

Order Now

Specification Test Results

Appearance Clear colorless to yellow Viscous liquid or low melting solid
Infrared spectrum Conforms
Refractive index 1.4720 to 1.4750 (20°C, 589 nm)
Specific optical rotation -71° to -74° (20°C, 589 nm) (c=2, CH3OH)
Titration with NaOH 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR