Ethylboronic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

37086-0010 37086-0050
TS37086-0010EA 45 USD
TS37086-0010 TS37086-0050
Ethylboronic acid 95%
Ethylboronic acid
Formula: C₂H₇BO₂
Storage Temperature: Refrigerator
MDL Number: MFCD01074536
CAS Number: 4433-63-0

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or flakes or needles
Infrared spectrum Conforms
Titration with NaOH ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR