Dimethyl bromomalonate 90% tech.
Supplier: Thermo Scientific Chemicals
|
Danger
|
30519-0050
30519-0250
TS30519-0050EA
51
USD
TS30519-0050
TS30519-0250
Dimethyl bromomalonate 90% tech.
Dimethyl bromomalonate
Formula:
C₅H₇BrO₄ MW: 211.01 g/mol Boiling Pt: 105…108 °C (11 mmHg) Density: 1.6 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00025865 CAS Number: 868-26-8 EINECS: 212-775-4 UN: 1760 ADR: 8,III |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4580 to 1.4660 (20°C, 589 nm) |
GC | ≥88 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...