Diethyl ethoxymethylenemalonate 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Diethyl 2-(ethoxymethylene)malonate

11434-1000 11434-5000
TS11434-1000EA 36 USD
TS11434-1000 TS11434-5000
Diethyl ethoxymethylenemalonate 99+%
Diethyl ethoxymethylenemalonate
Formula: C₁₀H₁₆O₅
MW: 216.23 g/mol
Density: 1.08 g/cm³
Flash Pt: 144 °C
Storage Temperature: Ambient
MDL Number: MFCD00009148
CAS Number: 87-13-8
EINECS: 201-725-7

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4610 to 1.4630 (20°C, 589 nm)
GC ≤1 % (ethanol)
Color scale ≤20 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR