Methyl isobutyrate 99%

Supplier: Thermo Scientific Chemicals
Danger

16460-1000 16460-5000
TS16460-1000EA 41 USD
TS16460-1000 TS16460-5000
Methyl isobutyrate 99%
Methyl isobutyrate
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Melting Pt: 85…84 °C
Density: 0.89 g/cm³
Flash Pt: 1 °C
Storage Temperature: Ambient
MDL Number: MFCD00008914
CAS Number: 547-63-7
EINECS: 208-929-5
UN: 3272
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3820 to 1.3860 (20°C, 589 nm)
Density (20°C) 0.886 to 0.892 g/ml
GC ≥98.5 %
Acid value ≤1 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR