Oxamide 98%

Supplier: Thermo Scientific Chemicals
Warning

12965-1000 12965-5000
TS12965-1000EA 48 USD
TS12965-1000 TS12965-5000
Oxamide 98%
Oxamide
Formula: C₂H₄N₂O₂
MW: 88.07 g/mol
Melting Pt: 491 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008007
CAS Number: 471-46-5
EINECS: 207-442-5

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration excess NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR