2,4-Hexadiene mixture of isomers 90% tech.

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hexa-2,4-diene

34667-0010 34667-0050
TS34667-0010EA 45 USD
TS34667-0010 TS34667-0050
2,4-Hexadiene mixture of isomers 90% tech.
2,4-Hexadiene
Formula: C₆H₁₀
MW: 82.15 g/mol
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: –79 °C
Density: 0.72 g/cm³
Flash Pt: –8 °C
MDL Number: MFCD00009297
CAS Number: 592-46-1
EINECS: 209-757-3
UN: 2458
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
GC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR