Methyl benzenesulphonate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
41399-0250
41399-1000
TS41399-0250EA
55
USD
TS41399-0250
TS41399-1000
Methyl benzenesulphonate 99%
Methyl benzenesulphonate
Formula:
C₇H₈O₃S Boiling Pt: 280 °C Melting Pt: –4 °C Density: 1.3 g/cm³ Flash Pt: 143 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014737 CAS Number: 80-18-2 EINECS: 201-256-8 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to light brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5150 to 1.5180 (20°C, 589 nm) |
HPLC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...