Trimesic acid trimethyl ester 99%

Supplier: Thermo Scientific Chemicals

15207-0250 15207-1000
TS15207-0250EA 98 USD
TS15207-0250 TS15207-1000
Trimesic acid trimethyl ester 99%
Trimesic acid trimethyl ester
Formula: C₁₂H₁₂O₆
Melting Pt: 144…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00008434
CAS Number: 2672-58-4
EINECS: 220-215-5

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 143 to 147 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR