(3-Pyridyl)acetic acid ethyl ester 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 3-pyridineacetate, Ethyl 2-(pyridin-3-yl)acetate, Ethyl (3-pyridyl)acetate, Ethyl 3-pyridylacetate

11892-0050 11892-0250
TS11892-0050EA 70 USD
TS11892-0050 TS11892-0250
(3-Pyridyl)acetic acid ethyl ester 99%
(3-Pyridyl)acetic acid ethyl ester
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 80 °C (.4 mmHg)
Density: 1.08 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006411
CAS Number: 39931-77-6
EINECS: 254-707-6

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4990 to 1.5010 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR