3-Nitrobenzamide 98%

Supplier: Thermo Scientific Chemicals
Warning

18344-0250 18344-1000
TS18344-0250EA 56 USD
TS18344-0250 TS18344-1000
3-Nitrobenzamide 98%
3-Nitrobenzamide
Formula: C₇H₆N₂O₃
MW: 166.14 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 140…144 °C
MDL Number: MFCD00007984
CAS Number: 645-09-0
EINECS: 211-431-0

Order Now

Specification Test Results

Appearance Yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 140 to 144 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR