(1S)-(+)-Camphor-10-sulfonic acid 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
((1S,4R)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulphonic acid, (1S)-(+)-2-Oxobornane-10-sulphonic acid, D(+)-Camphor-10-sulphonic acid, (1S)-(+)-7,7-Dimethylbicyclo(2.2.1)-2-heptanon-1-methane sulphonic acid, (1S)-(+)-10-Camphorsulphonic acid, D-Camphorsulphonic acid
10822-1000
10822-5000
TS10822-1000EA
82
USD
TS10822-1000
TS10822-5000
(1S)-(+)-Camphor-10-sulfonic acid 99%
(1S)-(+)-Camphor-10-sulphonic acid
Formula:
C₁₀H₁₆O₄S MW: 232.3 g/mol Boiling Pt: 375 °C (1013 hPa) Melting Pt: >190 °C (dec.) |
MDL Number:
MFCD00064157 CAS Number: 3144-16-9 EINECS: 221-554-1 UN: 3261 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | ≥190 °C |
Loss on drying | ≤2 % (1 g, 105 °C, 5 hrs) |
Specific optical rotation | +21° to +23° (25 °C, 589 nm) (c=20, H2O) |
Titration with NaOH | 98.5 to 101.5 % (On dried substance) |
Sulfate (SO4) | ≤500 ppm |
Heavy metals (as Pb) | ≤20 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...