Butyl (±)-lactate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Butyl 2-hydroxypropanoate, n-Butyl lactate, Butyl (±)-2-hydroxypropionate, (±)-Butyl lactate

25018-0025 25018-5000
TS25018-0025EA 357 USD
TS25018-0025 TS25018-5000
Butyl (±)-lactate 99%
Butyl (±)-lactate
Formula: C₇H₁₄O₃
MW: 146.19 g/mol
Boiling Pt: 185…187 °C (760 mmHg)
Melting Pt: –43 °C
Density: 0.98 g/cm³
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00004519
CAS Number: 138-22-7
EINECS: 205-316-4

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4200 to 1.4220 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR