3-Tolylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

34442-0010 34442-0050
TS34442-0010EA 29 USD
TS34442-0010 TS34442-0050
3-Tolylboronic acid 97%
3-Tolylboronic acid
Formula: C₇H₉BO₂
Melting Pt: 160…162 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00040198
CAS Number: 17933-03-8

Order Now

Specification Test Results

Appearance White to cream to beige Powder and/or chunks
Infrared spectrum Conforms
Melting point 159 to 165 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR