3-Aminophenylboronic acid monohydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dihydroxy(3-aminophenyl)borane monohydrate, 3-Aminobenzeneboronic acid monohydrate, 3-Aminobenzeneboronic acid, 3-Aminophenylboronic Acid Monohydrate, (3-Aminophenyl)metaboric acid monohydrate

35276-0010 35276-0050
TS35276-0010EA 62 USD
TS35276-0010 TS35276-0050
3-Aminophenylboronic acid monohydrate 98%
3-Aminophenylboronic acid monohydrate
Formula: C₆H₈BNO₂·1H₂O
MW: 154.96 g/mol
Melting Pt: 94…97 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00149554
CAS Number: 206658-89-1
EINECS: 250-189-0

Order Now

Specification Test Results

Appearance Beige to brown Crystalline powder or crystals
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR