2-Tolylboronic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

30926-0010 30926-0050
TS30926-0010EA 35 USD
TS30926-0010 TS30926-0050
2-Tolylboronic acid 95%
2-Tolylboronic acid
Formula: C₇H₉BO₂
Melting Pt: 164…165 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00093526
CAS Number: 16419-60-6

Order Now

Specification Test Results

Appearance White to beige Powder or crystals
Infrared spectrum Conforms
Melting point 160 to 166 °C
Additional info May contain varying amounts of anhydrides
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR