Ethyl crotonate 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: trans-Crotonic acid ethyl ester, 2-Butenoic acid ethyl ester, Crotonic acid ethyl ester, Ethyl trans-2-butenoate, trans-Ethyl crotonate, trans-2-Butenoic acid ethyl ester

16138-1000 16138-5000
TS16138-1000EA 39 USD
TS16138-1000 TS16138-5000
Ethyl crotonate 96%
Ethyl crotonate
Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 143 °C (1013 hPa)
Melting Pt: –79 °C
Density: 0.918 g/cm³
Flash Pt: 34 °C
MDL Number: MFCD00009289
CAS Number: 623-70-1
EINECS: 210-808-7
UN: 1862
ADR: 3,II
Merck Index: 13,02625

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4220 to 1.4280 (20°C, 589 nm)
Density (20/4°C) 0.916 to 0.921 g/ml
GC ≥95 %
Acid value ≤1 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR