Methyl phenylpropiolate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl Phenylpropiolate, Phenylpropiolic acid methyl ester

33459-0050 33459-0250
TS33459-0050EA 74 USD
TS33459-0050 TS33459-0250
Methyl phenylpropiolate 98%
Methyl phenylpropiolate
Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Boiling Pt: 109…110 °C (2 mmHg)
Melting Pt: 26 °C
MDL Number: MFCD00041685
CAS Number: 4891-38-7

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5615 to 1.5645 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR