Ethyl phenylpropiolate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Phenylpropiolic acid ethyl ester, Ethyl 3-phenylprop-2-ynoate, Ethyl phenylacetylenecarboxylate

11885-0050 11885-0250
TS11885-0050EA 42 USD
TS11885-0050 TS11885-0250
Ethyl phenylpropiolate 98%
Ethyl phenylpropiolate
Formula: C₁₁H₁₀O₂
MW: 174.2 g/mol
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 60 °C
Density: 1.05 g/cm³
Flash Pt: >112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009185
CAS Number: 2216-94-6
EINECS: 218-703-8

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5510 to 1.5530 (20°C, 589 nm)
GC ≥97.5 %
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR