Phenylpropiolic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Phenylacetylenecarboxylic acid, Phenylpropynoic acid, 3-Phenylpropiolic acid

13085-0050 13085-0250
TS13085-0050EA 68 USD
TS13085-0050 TS13085-0250
Phenylpropiolic acid 97%
Phenylpropiolic acid
Formula: C₉H₆O₂
MW: 146.15 g/mol
Boiling Pt: 294 °C (1013 hPa)
Melting Pt: 136…139 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004361
CAS Number: 637-44-5
EINECS: 211-285-8

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 136 to 139 °C
Titration with NaOH ≥96.0 %
UV at 270 nm A: ≤0.005
Color scale ≤10 APHA
Residue after evaporation ≤2 ppm
Acidity/alkalinity ≤0.0003 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR