Methyl dichloroacetate 99%

Supplier: Thermo Scientific Chemicals
Warning

14862-1000 14862-5000
TS14862-1000EA 26 USD
TS14862-1000 TS14862-5000
Methyl dichloroacetate 99%
Methyl dichloroacetate
Formula: C₃H₄Cl₂O₂
MW: 142.97 g/mol
Boiling Pt: 143 °C (1013 hPa)
Melting Pt: –52 °C
Density: 1.381 g/cm³
Flash Pt: 80 °C
MDL Number: MFCD00000843
CAS Number: 116-54-1
EINECS: 204-146-8
UN: 2299
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4411 to 1.4431 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR