Ethyl levulinate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Ethyl 4-oxopentanoate, 4-Oxovaleric acid ethyl ester, Ethyl 4-oxovalerate, 4-Ketovaleric acid ethyl ester, 4-Oxopentanoic acid ethyl ester, Levulinic acid ethyl ester
15362-1000
15362-5000
TS15362-1000EA
50
USD
TS15362-1000
TS15362-5000
Ethyl levulinate 98%
Ethyl levulinate
Formula:
C₇H₁₂O₃ Boiling Pt: 93 °C (1013 hPa) Density: 1.012 g/cm³ Flash Pt: 90 °C |
MDL Number:
MFCD00009209 CAS Number: 539-88-8 EINECS: 208-728-2 Merck Index: 13,03852 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4210 to 1.4240 (20°C, 589 nm) |
Density | (20°C) 1.009 to 1.014 g/ml |
GC | ≥97.5 % |
Acid value | ≤5 mg KOH/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...