Ethyl isovalerate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl isopentanoate, Isovaleric acid ethyl ester, 3-Methylbutyric acid ethyl ester, Isopropyl acetic acid ethyl ester, 3-Methylbutanoic acid ethyl ester

15048-0050 15048-1000
TS15048-0050EA 27 USD
TS15048-0050 TS15048-1000
Ethyl isovalerate 99%
Ethyl isovalerate
Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 131…133 °C (760 mmHg)
Melting Pt: –99 °C
Density: 0.863 g/cm³
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00009203
CAS Number: 108-64-5
EINECS: 203-602-3
UN: 1993
ADR: 3,III
Merck Index: 13,03849

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3950 to 1.3980 (20°C, 589 nm)
GC ≥98.5 %
Specific gravity (25/25°C) 0.861 to 0.865

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR