3-(4-Hydroxyphenyl)propionic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
12175-0050
12175-0250
12175-1000
TS12175-0050EA
33
USD
TS12175-0050
TS12175-0250
TS12175-1000
3-(4-Hydroxyphenyl)propionic acid 98%
3-(4-Hydroxyphenyl)propionic acid
Formula:
C₉H₁₀O₃ Melting Pt: 126…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002778 CAS Number: 501-97-3 EINECS: 207-931-3 |
Specification Test Results
Appearance | Off-white to beige Powder or granules |
Infrared spectrum | Conforms |
Melting point | 126 to 130 °C |
GC | ≥97.5 % |
Titration with NaOH | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...