Disodium succinate, anhydrous 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,4-Butanedioic acid disodium salt, Sodium Succinate, Succinic acid disodium salt, Sodium succinate dibasic
20874-1000
20874-5000
TS20874-1000EA
28
USD
TS20874-1000
TS20874-5000
Disodium succinate, anhydrous 99%
Disodium succinate
Formula:
C₄H₄Na₂O₄ MW: 162.05 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002790 CAS Number: 150-90-3 EINECS: 205-778-7 Merck Index: 12,08828 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤0.3 % (120°C) |
Titration with HClO4 | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...