N-(Hydroxymethyl)phthalimide 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(Hydroxymethyl)isoindoline-1,3-dione

12149-0250 12149-1000
TS12149-0250EA 30 USD
TS12149-0250 TS12149-1000
N-(Hydroxymethyl)phthalimide 97%
N-(Hydroxymethyl)phthalimide
Formula: C₉H₇NO₃
MW: 177.16 g/mol
Boiling Pt: 415 °C (1013 hPa)
Melting Pt: 140…145 °C
MDL Number: MFCD00005899
CAS Number: 118-29-6
EINECS: 204-241-4

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 140 to 145 °C
Loss on drying ≤2 %
Sulfated ash ≤0.1 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR