N-(3-Hydroxypropyl)phthalimide 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(3-Hydroxypropyl)isoindoline-1,3-dione

37701-0250 37701-1000
TS37701-0250EA 49 USD
TS37701-0250 TS37701-1000
N-(3-Hydroxypropyl)phthalimide 99%
N-(3-Hydroxypropyl)phthalimide
Formula: C₁₁H₁₁NO₃
Melting Pt: 75…77 °C
MDL Number: MFCD00023097
CAS Number: 883-44-3
EINECS: 212-931-1

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 75 to 78 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR