N-(3-Bromopropyl)phthalimide 98%

Supplier: Thermo Scientific Chemicals

10739-0250 10739-1000
TS10739-0250EA 46 USD
TS10739-0250 TS10739-1000
N-(3-Bromopropyl)phthalimide 98%
N-(3-Bromopropyl)phthalimide
Formula: C₁₁H₁₀BrNO₂
Boiling Pt: 433 °C (1013 hPa)
Melting Pt: 70…76 °C
MDL Number: MFCD00005904
CAS Number: 5460-29-7
EINECS: 226-738-5

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 70 to 76 °C
HPLC ≥97.5 %
Iron (Fe) ≤100 ppm
Heavy metals (as Pb) ≤50 ppm
Nitrilotriacetic acid ≤0.1 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR