N-(2-Bromoethyl)phthalimide 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(2-Bromoethyl)isoindoline-1,3-dione

10697-0250 10697-1000
TS10697-0250EA 49 USD
TS10697-0250 TS10697-1000
N-(2-Bromoethyl)phthalimide 97%
N-(2-Bromoethyl)phthalimide
Formula: C₁₀H₈BrNO₂
MW: 254.08 g/mol
Melting Pt: 81…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00005902
CAS Number: 574-98-1
EINECS: 209-379-9

Order Now

Specification Test Results

Appearance White to beige to light pink Crystalline powder
Infrared spectrum Conforms
Melting point 81 to 84 °C
Water ≤1 % (K.F.)
HPLC ≥96.0 % (on dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR