1,3-Dimethylurea 98%

Supplier: Thermo Scientific Chemicals

Synonyms: N,N'-Dimethylurea

11691-0010 11691-1000
TS11691-0010EA 68 USD
TS11691-0010 TS11691-0050 TS11691-1000
1,3-Dimethylurea 98%
1,3-Dimethylurea
Formula: C₃H₈N₂O
MW: 88.11 g/mol
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 101…105 °C
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00008286
CAS Number: 96-31-1
EINECS: 202-498-7

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 103 to 108 °C
HPLC ≥97.5 %
Carboniz. sub. by hot H2SO4 (tartrates, etc) Passes test
Chloride (Cl) ≤10 ppm
Oxalate (C2O4) (limit about 0.05%) Passes test
Phosphate (PO4) ≤10 ppm
Iron (Fe) ≤3 ppm
Lead (Pb) ≤2 ppm
Sulfur compounds (as SO4) ≤20 ppm
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR