Methyl (R)-(+)-lactate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl D-(+)-Lactate, (R)-Methyl 2-hydroxypropanoate, D-(+)-Lactic acid methyll ester, Methyl (R)-(+)-2-hydroxypropionate
41449-0050
41449-0250
TS41449-0050EA
75
USD
TS41449-0050
TS41449-0250
Methyl (R)-(+)-lactate 98%
Methyl (R)-(+)-lactate
Formula:
C₄H₈O₃ MW: 104.11 g/mol Boiling Pt: 144 Density: 1.09 g/cm³ Flash Pt: 49 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004517 CAS Number: 17392-83-5 EINECS: 241-420-6 UN: 3272 ADR: 3 III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4110 to 1.4150 (20°C, 589 nm) |
GC | ≥97.5 % |
Optical rotation | +6.5° to +10.5° (20°C, 589 nm) (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...