meso-2,3-Dimercaptosuccinic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

22667-0050 22667-0250 22667-1000
TS22667-0050EA 85 USD
TS22667-0050 TS22667-0250 TS22667-1000
meso-2,3-Dimercaptosuccinic acid 98%
meso-2,3-Dimercaptosuccinic acid
Formula: C₄H₆O₄S₂
Melting Pt: 196…198 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD00064799
CAS Number: 304-55-2
EINECS: 206-155-2
Merck Index: 12,09034

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤1 % (1 g, 105°C, 3 hrs)
Sulfated ash ≤0.2 %
Titration with NaOH ≥97.5 %
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR