(±)-Ethyl-2-methylbutyrate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (±)-2-Methylbutanoic acid ethyl ester, (±)-2-Methylbutyric acid ethyl ester, Ethyl 2-methylbutanoate

25026-0050 25026-1000
TS25026-0050EA 26 USD
TS25026-0050 TS25026-1000
(±)-Ethyl-2-methylbutyrate 99%
(±)-Ethyl-2-methylbutyrate
Formula: C₇H₁₄O₂
MDL Number: MFCD00012217
CAS Number: 7452-79-1

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3960 to 1.3990 (20°C, 589 nm)
Density (20/4°C) 0.864 to 0.871 g/ml
GC ≥98.5 %
Acid value ≤1 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR