Gibberellic acid 90%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: GA3, Gibberellin A3, (3S,3aS,4S,4aS,6S,8aR,8bR,11S)-6,11-Dihydroxy-3-methyl-12-methylene-2-oxo-4a,6-ethano-3,8b-prop-1-enoperhydroindeno[1,2-b]furan-4-carboxylic acid

11986-0010 11986-0050 11986-0250
TS11986-0010EA 64 USD
TS11986-0010 TS11986-0050 TS11986-0250
Gibberellic acid 90%
Gibberellic acid
Formula: C₁₉H₂₂O₆
MW: 346.38 g/mol
Melting Pt: 227 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00079329
CAS Number: 77-06-5
EINECS: 201-001-0
Merck Index: 13,04430

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤3 % (100°C, 7 hrs) (vacuum)
HPLC ≥88 %
Specific optical rotation +75° to +90° (20°C, 589 nm) (c=10, EtOH)
Arsenic (As) ≤3 ppm
Lead (Pb) ≤10 ppm
Heavy metals (as Pb) ≤40 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR