1,3-Propanesultone 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,2-Oxathiolane 2,2-dioxide

13142-0010 13142-0250 13142-1000
TS13142-0010EA 840 USD
TS13142-0010 TS13142-0250 TS13142-1000
1,3-Propanesultone 97%
1,3-Propanesultone
Formula: C₃H₆O₃S
MW: 122.14 g/mol
Boiling Pt: 180 °C (30 mmHg)
Melting Pt: 30…33 °C
Density: 1.392 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005355
CAS Number: 1120-71-4
EINECS: 214-317-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light yellow Low melting solid or liquid
Infrared spectrum Conforms
Melting point 28 to 33 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR