Ethyl 2-bromoisobutyrate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ethyl α-bromoisobutyrate, Ethyl 2-bromo-2-methylpropionate, 2-Brom-2-methylpropionic acid ethyl ester

11813-0050 11813-5000
TS11813-0050EA 26 USD
TS11813-0050 TS11813-5000
Ethyl 2-bromoisobutyrate 98%
Ethyl 2-bromoisobutyrate
Formula: C₆H₁₁BrO₂
Boiling Pt: 65 °C (1013 hPa)
Density: 1.329 g/cm³
Flash Pt: 60 °C
MDL Number: MFCD00000123
CAS Number: 600-00-0
EINECS: 209-980-6
UN: 1993

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4430 to 1.4460 (20°C, 589 nm)
GC ≥97.5 %
Sodium (Na) 30.5 to 32.5 % (as Na2SO4)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR