1-Propanesulfonic acid 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Propane-1-sulphonic acid

41834-0050 41834-0250
TS41834-0050EA 123 USD
TS41834-0050 TS41834-0250
1-Propanesulfonic acid 95%
1-Propanesulphonic acid
Formula: C₃H₈O₃S
MW: 124.16 g/mol
Boiling Pt: 120…121 °C (0.2 mmHg)
Melting Pt: 8 °C
Density: 1.25 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00041889
CAS Number: 5284-66-2
EINECS: 226-114-2
UN: 2586
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear brown to dark brown Liquid
Infrared spectrum Conforms
Refractive index 1.4380 to 1.4410 (20°C, 589 nm)
Titration with NaOH ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR