Ethyl iodoacetate 98% stabilised

Supplier: Thermo Scientific Chemicals
Danger

22219-0100 22219-0500 22219-2500
TS22219-0100EA 67 USD
TS22219-0100 TS22219-0500 TS22219-2500
Ethyl iodoacetate 98% stabilised
Ethyl iodoacetate
Formula: C₄H₇IO₂
MW: 214 g/mol
Boiling Pt: 179
Density: 1.8 g/cm³
Flash Pt: 76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001081
CAS Number: 623-48-3
EINECS: 210-796-3
UN: 2810
ADR: 6.1 II

Order Now

Specification Test Results

Appearance Clear light yellow to orange Liquid
Infrared spectrum Conforms
Refractive index 1.5025 to 1.5055 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer copper


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR