Methyl carbamate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Carbamic acid methyl ester

26931-0010 26931-0050 26931-2500
TS26931-0010EA 159 USD
TS26931-0010 TS26931-0050 TS26931-2500
Methyl carbamate 99%
Methyl carbamate
Formula: C₂H₅NO₂
Boiling Pt: 176…177 °C (760 mmHg)
Melting Pt: 54…56 °C
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00007964
CAS Number: 598-55-0
EINECS: 209-939-2
Merck Index: 12,06115

Order Now

Specification Test Results

Appearance White Adhering crystals
Infrared spectrum Conforms
Melting point 53 to 56 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR