Urethane 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Carbamic acid ethyl ester, Ethyl carbamate

32554-0010 32554-0500 32554-2500
TS32554-0010EA 268 USD
TS32554-0010 TS32554-0500 TS32554-2500
Urethane 97%
Urethane
Formula: C₃H₇NO₂
MW: 89.09 g/mol
Boiling Pt: 182
Melting Pt: 47…51 °C
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00007966
CAS Number: 51-79-6
EINECS: 200-123-1
Merck Index: 12,10013

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 46 to 51 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR