tert-Butyl carbamate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Carbamic acid tert-butyl ester

17345-0010 17345-0050 17345-0250
TS17345-0010EA 26 USD
TS17345-0010 TS17345-0050 TS17345-0250
tert-Butyl carbamate 98%
tert-Butyl carbamate
Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
Melting Pt: 105…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00007962
CAS Number: 4248-19-5
EINECS: 224-209-3

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystals or needles
Infrared spectrum Conforms
Melting point 106 to 110 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR