Methyl isonicotinate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Pyridinecarboxylic acid methyl ester, 4-Picolinic acid methyl ester, Methyl 4-pyridinecarboxylate, Pyridine-4-carboxylic acid methyl ester, Isonicotinic acid methyl ester

12706-1000 12706-5000
TS12706-1000EA 77 USD
TS12706-1000 TS12706-5000
Methyl isonicotinate 98%
Methyl isonicotinate
Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 207 °C (1013 hPa)
Melting Pt: 8.5 °C
Density: 1.161 g/cm³
Flash Pt: 82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006427
CAS Number: 2459-09-8
EINECS: 219-546-8

Order Now

Specification Test Results

Appearance Clear orange to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5110 to 1.5140 (20°C, 589 nm)
GC ≤1.0 % Diethyl carbonate
Color scale ≤50 APHA
Free acid (HCl) ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR