Acenaphthene 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2-Dihydroacenaphthylene

20134-0050 20134-1000 20134-5000
TS20134-0050EA 33 USD
TS20134-0050 TS20134-1000 TS20134-5000
Acenaphthene 99%
Acenaphthene
Formula: C₁₂H₁₀
MW: 154.21 g/mol
Boiling Pt: 279 °C (1013 hPa)
Melting Pt: 90…95 °C
Density: 1.06 g/cm³
Flash Pt: 135 °C (open cup)
Storage Temperature: Ambient
MDL Number: MFCD00003807
CAS Number: 83-32-9
EINECS: 201-469-6
UN: 3077
ADR: 9,III
Merck Index: 13,00029
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5520bcac8d97ebc91a4b9c3873c734cd4abd6427&vid=0cd04594d640995040d469a56bffa409d582f3ce&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to yellow to brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 91 to 95 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR