Cyclopropanecarbonyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11163-0050
11163-0250
11163-1000
11163-5000
TS11163-0050EA
26
USD
TS11163-0050
TS11163-0250
TS11163-1000
TS11163-5000
Cyclopropanecarbonyl chloride 98%
Cyclopropanecarbonyl chloride
Formula:
C₄H₅ClO Boiling Pt: 119 °C (760 mmHg) Density: 1.15 g/cm³ Flash Pt: 23 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001277 CAS Number: 4023-34-1 EINECS: 223-684-4 UN: 2920 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4515 to 1.4535 (20°C, 589 nm) |
GC | ≥97.5 % |
Titration RCOCl | ≥97.5 % |
BSC Certification | Passes |
Microbial contamination | (Salmonella) Absent in 375g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...