Sodium toluene-4-sulfonate

Supplier: Thermo Scientific Chemicals
Warning

42123-1000 42123-5000
TS42123-1000EA 85 USD
TS42123-1000 TS42123-5000
Sodium toluene-4-sulfonate
Sodium toluene-4-sulphonate
Formula: C₇H₇NaO₃S
Melting Pt: >400 °C
Flash Pt: >500 °C
MDL Number: MFCD00798566
CAS Number: 657-84-1
EINECS: 211-522-5

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration after Ion exchange ≥97.5 %
Titration with TBAH ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR