Diphenylacetylene 99%

Supplier: Thermo Scientific Chemicals

11718-0050 11718-0250 11718-1000
TS11718-0050EA 38 USD
TS11718-0050 TS11718-0250 TS11718-1000
Diphenylacetylene 99%
Diphenylacetylene
Formula: C₁₄H₁₀
MW: 178.23 g/mol
Boiling Pt: 170 °C (19 mmHg)
Melting Pt: 58…61 °C
Density: 0.99 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00004786
CAS Number: 501-65-5
EINECS: 207-926-6
Merck Index: 12,09643

Order Now

Specification Test Results

Appearance Light yellow to brown Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 58 to 61 °C
GC ≥98.5 %
Calcium (Ca) ≤100 ppm
Magnesium (Mg) ≤50 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤500 ppm
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR