4-Hydroxybenzhydrazide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Hydroxybenzoic hydrazide, 4-Hydroxybenzohydrazide, p-Hydroxybenzoic acid hydrazide, 4-Hydroxybenzene-1-carbohydrazide, 4-Hydroxybenzoic acid hydrazide

12100-0250 12100-1000 12100-5000
TS12100-0250EA 61 USD
TS12100-0250 TS12100-1000 TS12100-5000
4-Hydroxybenzhydrazide 98%
4-Hydroxybenzhydrazide
Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Melting Pt: 260…264 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007605
CAS Number: 5351-23-5
EINECS: 226-326-5

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR