Benzhydrazide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzoylhydrazine, Benzoic acid hydrazide, Benzohydrazide, Benzoic hydrazide

10577-1000 10577-5000
TS10577-1000EA 82 USD
TS10577-1000 TS10577-5000
Benzhydrazide 98%
Benzhydrazide
Formula: C₇H₈N₂O
MW: 136.15 g/mol
Melting Pt: 113…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00007596
CAS Number: 613-94-5
EINECS: 210-363-9

Order Now

Specification Test Results

Appearance White to beige Powder or crystals or needles
Infrared spectrum Conforms
Melting point 113 to 117 °C
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR