Pivalic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,2-Dimethylpropionic acid, Trimethylacetic acid

14002-0025 14002-0050 14002-1000 14002-5000
TS14002-0025EA 281 USD
TS14002-0025 TS14002-0050 TS14002-1000 TS14002-5000
Pivalic acid 99%
Pivalic acid
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 163…164 °C (760 mmHg)
Melting Pt: 33…35 °C
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00004194
CAS Number: 75-98-9
EINECS: 200-922-5
Merck Index: 12,07667

Order Now

Specification Test Results

Appearance White to off-white Low melting crystalline mass
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR